Human Recombinant TNFR-1 (from E. coli)
New Product
Tumor necrosis factor receptor type 1 (TNFR-1) is expressed in most tissues and is activated by soluble and membrane-bound tumor necrosis factor alpha (TNF-α). TNFR-1 activates NF-κB and MAPK pathways to induce inflammation, promote apoptotic cell death, inhibit tumorigenesis, and inhibit viral replication.
- High quality Low endotoxin Affordable
: For research use only.
- Source:
- Species:
- Environmentally Preferable:
Frequently Bought Together