Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IL-20 (from E. coli)
Human Recombinant IL-20 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant IL-20 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q9NYY1
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein/peptide name:
    IL-20
  • Purity:
    ≥95%
  • Molecular weight:
    17.7/35.3 kDa
  • Sequence:
    MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together