Order Entry
Canada
ContactUsLinkComponent
Human Recombinant MIG/CXCL9 (from E. coli)
Human Recombinant MIG/CXCL9 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant MIG/CXCL9 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay:Currently undetermined; ED50 ≤Currently undetermined; Bioactivity ≥NA
  • Gene ID:
    Q07325
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    CXCL9|MIG
  • Protein/peptide name:
    MIG/CXCL9
  • Purity:
    ≥95%
  • Molecular weight:
    11.7 kDa
  • Sequence:
    TPVVRKGRCS CISTNQGTIH LQSLKDLKQF APSPSCEKIE IIATLKNGVQ TCLNPDSADV KELIKKWEKQ VSQKKKQKNG KKHQKKKVLK VRKSQRSRQK KTT
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together