Human Recombinant I-TAC (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interferon-inducible T cell alpha chemoattractant (I-TAC), or CXCL11, is expressed at high levels in leukocytes, pancreas, and liver cells. I-TAC gene expression is induced by interferons alpha (IFN-α), beta (IFN-β), and gamma (IFN-ɣ). I-TAC is the dominant ligand known to bind the chemokine receptor CXCR3, thus acting as a stronge agonist. I-TAC functions as a chemoattractant for interleukin 2 (IL-2)-activated T cells.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: Currently undetermined; ED50 ≤Currently undetermined; Bioactivity ≥NA
- Gene ID:O14625
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CXCL11|B-R1
- Protein/peptide name:I-TAC
- Purity:≥95%
- Molecular weight:8.3 kDa
- Sequence:FPMFKRGRCL CIGPGVKAVK VADIEKASIM YPSNNCDKIE VIITLKENKG QRCLNPKSKQ ARLIIKKVER KNF
- Endotoxin level:Low
- Formulation:0.1% trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together