Human Recombinant MIP-4 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Macrophage inflammatory protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta, and bone marrow. MIP-4 receptors include the chemokine receptor 8 (CCR8), the G protein-coupled receptor 30 (GPR30), and the phosphatidylinositol transfer protein membrane-associate 3 (PITPNM3). MIP-4 acts as a chemoattractant for naive T cells, CD4+ T cells, CD8+ T cells, and nonactivated lymphocytes. Further, MIP-4 promotes breast cancer metastasis and attenuates the activation of acute lymphocytic leukemia B cells.
- High quality Low endotoxin Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P13501
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL18|PARC|AMAC-1|DC-CK2
- Protein/peptide name:MIP-4
- Purity:≥95%
- Molecular weight:7.9 kDa
- Sequence:AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together