Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant IL-2 (from E. coli)
New Product
 
undefined
Human Recombinant IL-2 (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay:CTLL-2 cell proliferation; ED50 ≤3 ng/ml; Bioactivity ≥3.3×10⁵ units/mg
  • Gene ID:
    P60568
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    T cell growth factor|TCGF|Aldesleukin
  • Protein/peptide name:
    IL-2
  • Purity:
    ≥97%
  • Molecular weight:
    15.5 kDa
  • Sequence:
    MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFCQSII STLT
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together