About this item
FG Cleanroom Saturix 3900SEQ wipes feature 100% polyester with sealed edges and advanced cleaning performance. Engineered for critical environments, they offer superior absorbency, strength, and ultra-low particle generation, ensuring optimal contamination control for cleanrooms, labs, and sensitive manufacturing processes.
2-ply, quilted construction
- Sealed edges
- Raised surface texture
- Pure, 100% continuous filament polyester fibers
- Laundered utilizing reverse osmosis and de-ionized water; packaged in ISO 4 cleanroom
- Extreme durability with versatile dry and wet wiping capabilities
- Low particle generation, extractables, and metallic ions
The FG Cleanwipes Saturix Poly Wipes are high-performance cleanroom wipes crafted from 100% polyester, designed for superior cleanliness, strength, and contamination control in critical environments. These wipes feature low-linting properties, excellent durability, and exceptional absorbency, making them ideal for wiping surfaces, absorbing spills, and removing contaminants without shedding fibers. Their advanced design ensures minimal particulate release, supporting cleanroom compliance and product integrity. The FG Saturix Poly Wipes provide a reliable, cost-effective solution for industries that demand the highest standards of cleanliness, durability, and operational efficiency.
Target industries include:
Pharmaceutical and Biotech: ideal for sterile manufacturing, surface wiping, and contamination control.
Electronics and Semiconductor: ensures pristine, particulate-free surfaces during assembly and precision cleaning.
Medical Device Manufacturing: supports stringent cleanliness standards for cleanrooms and production areas.
Aerospace and Automotive: effective for maintaining contaminant-free environments in sensitive manufacturing processes.