Palm Coated Polyurethane Industrial Gloves with HDPE Liner
Supplier: MAPA
New Product
Certificates
About this item
Standard protection for intricate handling in reasonably clean environments.
Great dexterity
Maximum comfort
ANSI cut level A2
13 gauge HDPE liner
Smooth grip
Knit wrist
These gloves are designed for use in industrial environments.
Specifications
- Catalog No:
- 77868-144
- 77868-146
- 77868-148
- 77868-150
- 77868-152
- 77868-154
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- PU
- PU
- PU
- PU
- PU
- PU
- Finish:
- Smooth grip
- Smooth grip
- Smooth grip
- Smooth grip
- Smooth grip
- Smooth grip
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile