Order Entry
Canada
Orders LinkContactUsLinkComponent
Abgene™ Sample Processing and Storage Plates

Abgene™ Sample Processing and Storage Plates

Abgene™ Sample Processing and Storage Plates
Certificates
Abgene™ Sample Processing and Storage Plates
Abgene™ Sample Processing and Storage Plates

14 Options Available Below

Product Details & Documents

Abgene™ polypropylene MicroWell and DeepWell plates provide performance and value for sample storage, high-throughput screening, and molecular diagnostic assays. Abgene™ plates are made with high-quality, medical-grade, virgin polypropylene resin. These plates benefit from exceptional design and manufacturing advancements that ensure sample integrity and facilitate workflow efficiency.

  • Made of virgin, medical-grade polypropylene

  • hDNA-free, DNase-/RNase-free, and PCR inhibitor-free
  • Low extractables and leachables
  • 48 to 384 well plate formats available
  • Temperature range: −80 to +121 °C
  • Volume range: 55 µl to 6 ml
  • ANSI/SLAS-compliant

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries, or storing samples for either intermediate or long-term use. Abgene™ microtiter plates are manufactured to exacting specifications in Class 100000 cleanroom ISO 9001 conditions using high-quality medical grade virgin USP <661.1> polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handlers for high-throughput workflows. These polypropylene storage plates are offered with spindle, pyramidal, U, and V bottom wells along with a wide range of volumes to accommodate most applications.

Packaging: Abgene™ Storage plates are available individually or bulk packed.

Product Family Options

48-well plates

Product Information

  • Total Volume
    No. of wells
    Bottom style
    Color
    Sterility
    Well shape
    Availability
    Price
  • 77753-710
    Storage plate
    Storage plate
    6000 µl
    48
    Pyramidal
    Natural
    Cleanroom-manufactured
    Rectangular
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $712.23
    $712.23
    New Customer Promotional Price
    Case of 50
       $712.23

    Supplier Number: AB-0988

96-well plates

Product Information

  • Total Volume
    No. of wells
    Bottom style
    Color
    Sterility
    Well shape
    Availability
    Price
  • 77753-702
    Storage plate (Individual)
    Storage plate (Individual)
    800 µl
    96
    V-conical
    Natural
    Cleanroom-manufactured
    Round
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $425.42
    $425.42
    New Customer Promotional Price
    Case of 50
       $425.42

    Supplier Number: AB-0765
  • 77753-708
    Storage plate (Bulk)
    Storage plate (Bulk)
    1200 µl
    96
    U-bottom
    Natural
    Cleanroom-manufactured
    Round
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $559.88
    $559.88
    New Customer Promotional Price
    Case of 50
       $559.88

    Supplier Number: AB-0787
  • 77753-674
    Storage plate (Bulk)
    Storage plate (Bulk)
    2200 µl
    96
    U-bottom
    Natural
    Cleanroom-manufactured
    Square
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $611.44
    $611.44
    New Customer Promotional Price
    Case of 50
       $611.44

    Supplier Number: AB-0788
  • 77753-660
    Storage plate (Bulk)
    Storage plate (Bulk)
    330 µl
    96
    U-bottom
    Natural
    Cleanroom-manufactured
    Round
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $430.91
    $430.91
    New Customer Promotional Price
    Case of 100
       $430.91

    Supplier Number: AB-0796
  • 77753-652
    Storage plate (Bulk)
    Storage plate (Bulk)
    800 µl
    96
    V-conical
    Natural
    Cleanroom-manufactured
    Round
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $425.42
    $425.42
    New Customer Promotional Price
    Case of 50
       $425.42

    Supplier Number: AB-0859
  • 77753-676
    Storage plate (Individual)
    Storage plate (Individual)
    2200 µl
    96
    V-conical
    Natural
    Cleanroom-manufactured
    Square
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $611.44
    $611.44
    New Customer Promotional Price
    Case of 50
       $611.44

    Supplier Number: AB-0932
  • 77753-682
    Storage plate
    Storage plate
    200 µl
    96
    V-spindle
    Natural
    Cleanroom-manufactured
    Round
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $681.33
    $681.33
    New Customer Promotional Price
    Case of 100
       $681.33

    Supplier Number: AB-1058
  • 77753-680
    Storage plate
    Storage plate
    1200 µl
    96
    U-bottom
    Natural
    Cleanroom-manufactured
    Square
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $577.71
    $577.71
    New Customer Promotional Price
    Case of 50
       $577.71

    Supplier Number: AB-1127
  • 77753-698
    Storage plate (Individual)
    Storage plate (Individual)
    1200 µl
    96
    U-bottom
    Natural
    Cleanroom-manufactured
    Round
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $550.81
    $550.81
    New Customer Promotional Price
    Case of 50
       $550.81

    Supplier Number: AB-0564
  • 77753-684
    Storage plate (Individual)
    Storage plate (Individual)
    2200 µl
    96
    U-bottom
    Natural
    Cleanroom-manufactured
    Square
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $611.44
    $611.44
    New Customer Promotional Price
    Case of 50
       $611.44

    Supplier Number: AB-0661

384-well plates

Product Information

  • Total Volume
    No. of wells
    Bottom style
    Color
    Sterility
    Well shape
    Availability
    Price
  • 77753-650
    Storage plate
    Storage plate
    250 µl
    384
    V-conical
    Natural
    Cleanroom-manufactured
    Square
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $523.93
    $523.93
    New Customer Promotional Price
    Case of 50
       $523.93

    Supplier Number: AB-1178
  • 77753-686
    Storage plate
    Storage plate
    120 µl
    384
    Pyramidal
    Natural
    Cleanroom-manufactured
    Square
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $652.89
    $652.89
    New Customer Promotional Price
    Case of 50
       $652.89

    Supplier Number: AB-0781
  • 77753-654
    Storage plate
    Storage plate
    55 µl
    384
    V-spindle
    Natural
    Cleanroom-manufactured
    Round
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $635.58
    $635.58
    New Customer Promotional Price
    Case of 50
       $635.58

    Supplier Number: AB-1056
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.