VWR® Conductivity Electrodes, Avantor
Supplier: VWR International
New Product
Certificates
About this item
The VWR® Conductivity electrodes allow for the measurement of conductivity, TDS (total dissolved solids) resisitivity and salinity of the solution (dependent on the meter capabilities).
Variety of cell constants available to fit sample requirements
- Built in temperature probe to allow for ATC (automatic temperature compensation)
- Five pin aviation connection
- For use with VWR® Conductivity meters
For application specific recommendations, please reference 'VWR® Electrode Selection Guide’ located in the Documents section.
Frequently Bought Together