About this item
The VWR-DO950 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation.
High resolution color touch display (7.0")
- Air-saturated water or zero oxygen calibration capabilities
- Automatic or manual temperature compensation capabilities
- Auto-read, timed-read, and continuous-read measurement options
- Data capacity of 1000 sets for each parameter (GLP-compliant)
- Auto barometric pressure compensation
- For use with DO1 Polarographic DO Electrode (77618-816)
The easy to use VWR-DO950 portable meters provide accurate dissolved oxygen measurements for accurate water quality testing. The intelligent operation system provides features including user managment, calibration management, electrode management, method management, and many other advanced capabilities. Connect the meter easily to the VWR Dot Matrix Printer (10799-132) to print measurement reports.
For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the Related Information section.
IP 54.
Three-year meter warranty.
All meters include: Electrode stand, In use cover, and touch screen pen.