Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant CRYBA4 (from E. coli)

Human Recombinant CRYBA4 (from E. coli)

100 µg
Catalog # ABCA_AB113143-100U
New Product
Supplier:   Abcam
Supplier #:  AB113143-100UG
Human Recombinant CRYBA4 (from E. coli)
Catalog # ABCA_AB113143-100U
Supplier:  Abcam
New Product
CAS Number:  
Human Recombinant CRYBA4 (from E. coli)
Human Recombinant CRYBA4 (from E. coli)

100 µg

CAS Number:  
Price
$913.00
Log in for availability
Each (100µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant Human CRYBA4 protein is a Human Full Length protein, in the 1 to 196 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Size:
    100 µg
  • Form:
    Liquid
  • Protein/peptide name:
    CRYBA4
  • Protein synonyms:
    Beta-crystallin A4|Beta-A4 crystallin|CRYBA4
  • Purity:
    >95%
  • Concentration:
    1 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCA_AB113143-100U
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0.58% Sodium chloride, 0.32% Tris HCl, 0.02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications