Human Recombinant SIT (from E. coli)
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recombinant human SIT protein is a human fragment protein, in the 62 to 196 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Size:10 µg
- Form:Liquid
- Protein/peptide name:SIT
- Protein synonyms:SIT|SIT1|Signaling threshold-regulating transmembrane adapter 1|SHP2-interacting transmembrane adapter protein|Suppression-inducing transmembrane adapter 1|gp30/40
- Purity:>90%
- Concentration:0.25 mg/ml
- Source:E. coli
- Biological activity:Standard
- Shipping temperature:20
- Cat. No.:ABCA_AB113580-10UG
- Protein/peptide type:Recombinant
- Storage conditions:Store at –20 °C
- Formulation:pH: 8, Constituents: 20% glycerol (glycerin, glycerine), 0.32% Tris HCl
- Conjugation:Unconjugated
- Tested applications:MS, SDS-PAGE
- Species:Human
- Animal-free:N
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



