Mouse TYRO3 ELISA Kit (DTK)
Catalog # ABCA_AB267577-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:TYRO3
- Description:Mouse TYRO3 ELISA kit (DTK)
- Size:96 tests
- Sample type:Plasma, Cell culture supernatant, Serum
- Detection method:Colorimetric
- Detection range:0.4 - 100 ng/ml
- Storage temperature:4 °C
- Sensitivity:0.4 ng/ml
- Regulatory status:RUO
Specifications
About this item
Mouse TYRO3 ELISA Kit (DTK) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TYRO3 (DTK) in Mouse in Plasma, Cell culture supernatant, Serum samples.