Pig Haptoglobin ELISA Kit
ABCA_AB205091-1X96
: Abcam
New Product
:
- Description:Pig Haptoglobin ELISA kit
- Size:96 tests
Pig Haptoglobin ELISA Kit is a sandwich (quantitative) ELISA kit for the measurement of Pig Haptoglobin in Pig in Citrate plasma, EDTA Plasma, Heparin Plasma, Serum samples.