Order Entry
Canada
ContactUsLinkComponent
Human IL-10 ELISPOT Kit (with un-coated plates)
Human IL-10 ELISPOT Kit (with un-coated plates)
Supplier:  Abcam
New Product
Certificates
undefined
Human IL-10 ELISPOT Kit (with un-coated plates)
Supplier:  Abcam
New Product

Specifications

  • Species reactivity:
    Human;Monkey
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Antigen name:
    IL-10
  • Storage temperature:
    +4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO

Specifications

Frequently Bought Together