Human Tissue Kallikrein (KLK1) ELISA Kit- Extracellular
Supplier: Abcam
New Product
Certificates
About this item
Human Tissue Kallikrein (KLK1) ELISA Kit- Extracellular is a single-wash 90-min Simplestep used to quantify Human Tissue Kallikrein (KLK1) with a sensitivity of 3.9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.
Frequently Bought Together