ContactUsLinkComponent
Human Neurofilament H ELISA Kit (200 kDa Neurofilament protein)
Human Neurofilament H ELISA Kit (200 kDa Neurofilament protein)
Catalog # ABCA_AB272199-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human Neurofilament H ELISA Kit (200 kDa Neurofilament protein)
Catalog # ABCA_AB272199-1X96
Supplier:  Abcam
Supplier Number:  AB272199-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Human Neurofilament H ELISA kit (200 kDa Neurofilament protein)
  • Size:
    96 tests
Specifications
About this item
Human Neurofilament H ELISA kit (200 kDa Neurofilament protein) is a single-wash 90-min Simplestep used to quantify Human Neurofilament H (200 kDa Neurofilament protein) with a sensitivity of 141 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.