Order Entry
Canada
ContactUsLinkComponent
Mouse CD93 ELISA Kit
Mouse CD93 ELISA Kit
Catalog # ABCA_AB233620-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Mouse CD93 ELISA Kit
Catalog # ABCA_AB233620-1X96
Supplier:  Abcam
Supplier Number:  AB233620-1X96TESTS
New Product
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    CD93
  • Description:
    Mouse CD93 ELISA kit
  • Size:
    96 tests
  • Sample type:
    Tissue Extracts, Heparin Plasma, Citrate plasma, Tissue Homogenate, Cell culture supernatant, Serum, EDTA Plasma
  • Detection method:
    Colorimetric
  • Time to Results:
    1 hr 30 min
  • Detection range:
    23.4 - 1500 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    3.4 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse CD93 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse CD93 with a sensitivity of 3.4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.