Order Entry
Canada
Orders LinkContactUsLinkComponent
Sample Diluent NBP

Sample Diluent NBP

Sample diluent NBP, Research use only (RUO), 20 ml
Catalog # ABCA_AB221826-20ML
New Product
Supplier:   Abcam
Supplier #:  AB221826-20ML
Sample Diluent NBP
Catalog # ABCA_AB221826-20ML
Supplier:  Abcam
New Product
CAS Number:  
Sample Diluent NBP
Sample Diluent NBP

Sample diluent NBP, Research use only (RUO), 20 ml

CAS Number:  
Price
$164.00
Log in for availability
Each (20ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sample Diluent NBP is part of the reagents, controls and accessories range.

Caution: RUO.

Specifications

Specifications

  • Description:
    Sample diluent NBP
  • Type:
    Research use only (RUO)
  • Volume:
    20 ml
  • Cat. No.:
    ABCA_AB221826-20ML
  • Storage temperature:
    4 °C

Specifications