Human IL-1 beta ELISpot Pair
ABCA_AB281985-10X9
: Abcam
New Product
:
- Description:Human IL-1 beta ELISpot pair
- Size:10×96 tests
Human IL-1 beta ELISpot Pair is a ELISpot Pair for the sensitive detection of Human IL-1 beta production in a single cell suspension.