Mouse JAM-A ELISA Kit (Junctional Adhesion Molecule)
ABCA_AB277080-1X96
: Abcam
New Product
:
- Description:Mouse JAM-A ELISA kit (Junctional Adhesion Molecule)
- Size:96 tests
Mouse JAM-A ELISA Kit (Junctional Adhesion Molecule) is a single-wash 90-min Simplestep used to quantify Mouse JAM-A (Junctional Adhesion Molecule) with a sensitivity of 2.92 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair