Human Interferon gamma ELISPOT Set
Supplier: Abcam
New Product
Certificates
About this item
Human Interferon gamma ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Interferon gamma production in a single cell suspension.
Specifications
- Species reactivity:Human
- Sample type:Suspension cells
- Antigen name:Interferon gamma
- Storage temperature:+4 ℃
- Processing time:Multiple steps
- Regulatory status:RUO
Specifications
Frequently Bought Together