ContactUsLinkComponent
Human Superoxide Dismutase 1 ELISA Kit
Human Superoxide Dismutase 1 ELISA Kit
Catalog # ABCA_AB309312-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human Superoxide Dismutase 1 ELISA Kit
Catalog # ABCA_AB309312-1X96
Supplier:  Abcam
Supplier Number:  AB309312-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Human Superoxide Dismutase 1 ELISA kit
  • Size:
    96 tests
Specifications
About this item
Human Superoxide Dismutase 1 ELISA kit is a cell-based (quantitative) ELISA kit for the measurement of Human Superoxide Dismutase 1 in Human in tissue extracts, cell lysate samples.