Mouse Galectin 1 ELISA Kit
Catalog # ABCA_AB119595-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:Galectin 1
- Description:Mouse Galectin 1 ELISA kit
- Size:96 tests
- Sample type:Plasma, Cell culture supernatant, Serum, Cell Lysate
- Detection method:Colorimetric
- Detection range:156 - 10000 pg/ml
- Storage temperature:4 °C
- Sensitivity:5 pg/ml
- Regulatory status:RUO
Specifications
About this item
Mouse Galectin 1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Galectin 1 in Mouse in Plasma, Cell culture supernatant, Serum, Cell Lysate samples.