Mouse Interferon gamma ELISPOT Set
: Abcam
New Product
Mouse Interferon gamma ELISPOT Set is a ELISPOT Set for the sensitive detection of Mouse Interferon gamma production in a single cell suspension.
- Species reactivity:Mouse
- Antigen name:Interferon gamma
- Storage temperature:4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
Frequently Bought Together