Complex I Enzyme Activity Dipstick Assay Kit
Catalog # ABCA_AB109720-48TE
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Complex I Enzyme activity dipstick assay kit
- Size:48 tests
- Protein:Complex I
- Species reactivity:Other, Human, Mouse, Rat
- Sample type:Cell culture extracts, Tissue, Cell culture supernatant
- Storage temperature:+4 ℃
- Processing time:Multiple steps
- Product type:Pre-coated
- Format:Pre-coated
Specifications
About this item
Complex I Enzyme Activity Dipstick Assay kit is an immunoassay kit for the detection of Complex I Enzyme Activity in Mouse, Rat, Cow, Human in cell culture extracts, tissue, cell culture supernatant samples.
Caution: RUO