Rat Interferon gamma ELISPOT Kit
Supplier: Abcam
New Product
Certificates
About this item
Rat Interferon gamma ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Rat Interferon gamma production in a single cell suspension.
Specifications
- Species reactivity:Rat
- Sample type:Suspension cells
- Antigen name:Interferon gamma
- Storage temperature:4 °C
- Processing time:Multiple steps
Specifications
Frequently Bought Together