ContactUsLinkComponent
Mouse IGFBP-1 ELISA Kit
Mouse IGFBP-1 ELISA Kit
Catalog # ABCA_AB213865-1X96
Supplier:  Abcam
New Product
CAS Number:  
Mouse IGFBP-1 ELISA Kit
Catalog # ABCA_AB213865-1X96
Supplier:  Abcam
Supplier Number:  AB213865-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Mouse IGFBP-1 ELISA kit
  • Size:
    96 tests
Specifications
About this item
Mouse IGFBP-1 ELISA Kit is a sandwich (quantitative) ELISA kit for the measurement of Mouse IGFBP-1 in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.