Blocking Buffer 10X
Catalog # ABCA_AB210904-50ML
Supplier: Abcam
New Product
CAS Number:
Specifications
- Concentration:10X
- Description:Blocking buffer
- Storage temperature:4 °C
Specifications
About this item
Blocking Buffer 10X is part of the reagents, controls and accessories range.
Caution:
RUO.