Orders LinkContactUsLinkComponent
Gentamicin Sulfate Solution
Liquid, 20 ml
Catalog # 77975-388
New Product
 :  9355-20ML
Gentamicin Sulfate Solution
Catalog # 77975-388
New Product
CAS Number:  
Gentamicin Sulfate Solution
Gentamicin Sulfate Solution
Liquid, 20 ml
CAS Number:  
Price
$275.21
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms. Also effective against various strains of mycoplasma. Recommended concentration for cell culture application: 1 ml/L medium.
  • Animal-component free
  • Chemically defined
  • GMP manufacture
 : Store at 15 to 30 °C. Shelf life: 24 months from date of manufacture.
Specifications
Specifications
  • Media Format:
    Liquid
  • Size:
    20 ml
  • Cat. No.:
    77975-388
  • L-Glutamine:
    No
  • Calcium:
    No
  • HEPES:
    No
  • Sodium pyruvate:
    No
  • Glucose level:
    Without
  • Sodium bicarbonate:
    No
  • Magnesium:
    No
  • Stable glutamine:
    No
  • Phenol red:
    No
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.