Order Entry
Canada
ContactUsLinkComponent
PBS 10X-Dulbecco's Phosphate Buffered Saline Solution
PBS 10X-Dulbecco's Phosphate Buffered Saline Solution
Catalog # 77975-374
New Product
CAS Number:  
undefined
PBS 10X-Dulbecco's Phosphate Buffered Saline Solution
Catalog # 77975-374
Supplier Number:  9242-500ML
New Product
CAS Number:  

Specifications

  • Description:
    PBS 10X-Dulbecco's phosphate buffered saline solution media base
  • Size:
    500 ml
  • Calcium:
    No
  • Sodium bicarbonate:
    No
  • Phenol red:
    No
  • Format:
    Liquid

Specifications

About this item

PBS 10X Dulbecco's Phosphate buffered saline solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.

  • Does not contain phenol red, sodium bicarbonate, or calcium and magnesium salts

A common diluent and rinsing solution, Phosphate Buffered Saline Solution (PBS) maintains cell culture media in the physiological pH range. PBS is also useful for reconstituting enzymes for tissue dissociation or lifting cells from substrates.

Packaging:

Store at 15 to 30 °C

Shelf life: 24 months from date of manufacture