BalanCD™ Anti-Clumping Supplement
Catalog # 77975-338
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
New Product
CAS Number:
Specifications
- Media Format:Liquid
- Size:50 ml
Specifications
About this item
Part of the BalanCD system of media and supplements, the anti-clumping supplement is an animal-component- and enzyme-free supplement to cell culture medium, designed to reduce cell aggregation for various cell types in suspension culture.
Animal component and enzyme-free
Designed to reduce cell aggregation for various cell types in suspension culture
Anti-clumping supplement is designed to reduce cell aggregation for various cell types in suspension culture. This supplement is animal component and enzyme-free. This supplement is not intended to dissociate adherent cells.
Packaging: Store at 2 to 8 °C. Shelf life: 2 years from date of manufacture.