Order Entry
Canada
ContactUsLinkComponent
Trypsin (1:250) 2.5% 10X Solution
Trypsin (1:250) 2.5% 10X Solution
Catalog # 77975-384
New Product
CAS Number:  
undefined
Trypsin (1:250) 2.5% 10X Solution
Catalog # 77975-384
Supplier Number:  9336-100ML
New Product
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Size:
    100 ml

Specifications

About this item

Trypsin solution 10X is an enzyme preparation, used to remove cells from the culture vessel surface into suspension. This formulation is a 2.5% solution of 1:250 trypsin in normal saline.

  • Animal-component free
  • Chemically defined
  • GMP manufacture

Trypsin is a proteolytic enzyme used to dissociate adherent cells from the surface of culture vessels.

Packaging: Store at ≤−10 °C. Shelf life: 24 months from date of manufacture.