Trypsin EDTA 1X Solution in HBSS
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
New Product
Certificates
About this item
Contains glucose and phenol red.
Animal-component free
Chemically defined GMP manufacture
Trypsin-EDTA is a combined reagent used to release adherent cells fromthe culture vessel surface into suspension. This formulation contains 0.5 g/L trypsin (1:250) and 0.2 g/L of EDTA in Hanks' balanced salt solution.
Packaging:
Store at ≤–10 °C
Shelf life: 24 months from date of manufacture
Frequently Bought Together