DME High Glucose without L-Glutamine
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
New Product
Certificates
About this item
Dulbecco's Modified Eagle's medium (DME) is a modification of Basal Medium Eagle (BME) and contains a fourfold increase of amino acids, vitamins, and nutrients to provide support for a wide variety of cell lines.
- Animal component-free
- Chemically defined
- GMP manufacture
With:
4500mg/L Glucose
3700mg/L Sodium bicarbonate
Phenol red
Without:
L-Glutamine - add 20 ml/L (200 mM)
Sodium pyruvate
Specifications
- Calcium:Yes
- Magnesium:Yes
- HEPES:No
- Sodium bicarbonate:Yes
- Sodium pyruvate:No
- Phenol red:Yes
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:High
- Media Base:DMEM - Dulbecco's Modified Eagle Medium
Specifications
Frequently Bought Together