Iscove's Modified Dulbecco's Medium
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Iscove's Modified Dulbecco's Medium (IMDM) is a modification of Dulbecco's Basal Medium Eagle (BME) that uses high glucose and pyruvate, additional amino acids, 25 mM HEPES, selenium, and other components. IMDM is designed for serum-free growth of hemopoietic cells in culture and supports the growth of lymphocytes and a variety of other hybrid cells.
- Animal component-free
- Chemically defined
- GMP manufacture
With:
25 mM HEPES buffer
3024 mg/L Sodium bicarbonate
Without:
Alpha Thioglycerol
L-Glutamine, add 20 ml/L (200 mM)
Specifications
Specifications
- Media Format:Liquid
- Size:500 ml
- Cat. No.:77975-309
- L-Glutamine:No
- Calcium:Yes
- HEPES:Yes
- Sodium pyruvate:Yes
- Glucose level:High
- Sodium bicarbonate:Yes
- Magnesium:Yes
- Stable glutamine:No
- Media Base:IMDM - Iscove's Modified Dulbecco's Medium
- Phenol red:Yes
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



