PRIME-XV T Cell Expansion XSFM
77975-331
New Product
:
- Media Format:Liquid
- Pack type:Bottle
- Size:1 L
- Calcium:No
- Magnesium:No
- HEPES:No
- Sodium bicarbonate:No
- Sodium pyruvate:No
- Phenol red:Yes
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:Without
- Media Base:Proprietary mixture for human primary cells
Xeno-free, serum-free medium for T cell expansion.
- Contains gentamicin for reduced contamination of culture components
PRIME-XV T Cell XSFM is a xeno-free and serum-free culture medium that supports the expansion and reactivation of human T cells, as well as the transition from preclinical to clinical applications for immunotherapy therapeutics. Designed and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management.
: FDA, Federal, and State registered - cGMP-compliant manufacture EN ISO 13485:2016 certified MDSAP certified Extensive QC testing including functionality, sterility (USP <71>), endotoxin (USP <85>), and mycoplasma (USP <63>) Drug Master Files (DMFs) filed with the FDA – available upon request.