Order Entry
Canada
ContactUsLinkComponent
MBAs QC Concentrate, Certified Reference Material
MBAs QC Concentrate, Certified Reference Material
Catalog # NSPEI-124
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
undefined
MBAs QC Concentrate, Certified Reference Material
Catalog # NSPEI-124
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  QCI-124
CAS Number:  

Specifications

  • Concentration:
    0.2 - 1 µg/ml
  • Description:
    MBAs QC concentrate, certified reference material 10.5 ml
  • Shipping conditions:
    Non-hazardous
  • Physical form:
    Liquid
  • Contents:
    Surfactants - MBAS

Specifications

About this item

Meet or exceed performance criteria with QC samples.

  • Manufactured for AA, ICP, ICP-MS, IC, XRF and other analytical instrumentation
  • For US EPA Methods 531.1, 531.2, and 632

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016.