Order Entry
Canada
ContactUsLinkComponent
Anti-TCPTP Rabbit Monoclonal Antibody [clone: EPR28199-34]
Anti-TCPTP Rabbit Monoclonal Antibody [clone: EPR28199-34]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-TCPTP Rabbit Monoclonal Antibody [clone: EPR28199-34]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB314497-100U
  • ABCA_AB314497-1MG
  • Antigen symbol:
  • TCPTP
  • TCPTP
  • Antigen name:
  • PTPT
  • PTPT
  • Antigen synonyms:
  • PTPN2|Tyrosine-protein phosphatase non-receptor type 2|T-cell protein-tyrosine phosphatase
  • PTPN2|Tyrosine-protein phosphatase non-receptor type 2|T-cell protein-tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR28199-34
  • EPR28199-34
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 48473 Da
  • 48473 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage temperature:
  • Store at 4 °C
  • Store at 4 °C
  • Shipping temperature:
  • 4 °C
  • 4 °C
  • Tested applications:
  • Flow Cyt (Intra), IHC-P
  • Flow Cyt (Intra), IHC-P

Specifications

Frequently Bought Together