Anti-Cytokeratin 3/CK-3 Mouse Monoclonal Antibody [clone: AE5]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [AE5] to Cytokeratin 3/CK-3.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cytokeratin 3/CK-3
Clonality: Monoclonal
Clone: AE5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB68260-1MG
- ABCA_AB68260-100UG
- Antigen symbol:
- Cytokeratin 3/CK-3
- Cytokeratin 3/CK-3
- Antigen name:
- 65 kDa cytokeratin
- 65 kDa cytokeratin
- Antigen synonyms:
- Cytokeratin-3|Keratin-3|Type-II keratin Kb3|CK-3|K3|KRT3
- Cytokeratin-3|Keratin-3|Type-II keratin Kb3|CK-3|K3|KRT3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- AE5
- AE5
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 64417 Da
- 64417 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS
- PBS
- Storage temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested applications:
- IHC-P
- IHC-P
Specifications
Frequently Bought Together