Anti-DDDDK epitope tag Rabbit Monoclonal Antibody [clone: EPR20018-251]
: Abcam
New Product
Rabbit monoclonal [EPR20018-251] to DDDDK tag (Binds to FLAG® tag sequence).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: DDDDK epitope tag
Clonality: Monoclonal
Clone: EPR20018-251
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- ABCA_AB205606-100U
- ABCA_AB205606-1ML
- Antigen symbol:
- DDDDK epitope tag
- DDDDK epitope tag
- Antigen name:
- DDDK
- DDDK
- Antigen synonyms:
- DYKDDYKDDDDK (also known as the FLAG搴 tag sequence)|DYKDDDDK epitope tag|DYKDDDDK tag|ECS epitope tag|ECS tag|Enterokinase Cleavage Site epitope tag|Enterokinase Cleavage Site tag|FLAG搴|FLAG搴 tag|ddk
- DYKDDYKDDDDK (also known as the FLAG搴 tag sequence)|DYKDDDDK epitope tag|DYKDDDDK tag|ECS epitope tag|ECS tag|Enterokinase Cleavage Site epitope tag|Enterokinase Cleavage Site tag|FLAG搴|FLAG搴 tag|ddk
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR20018-251
- EPR20018-251
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Frequently Bought Together