Anti-HIRA/HIR Rabbit Monoclonal Antibody [clone: EPR7416]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR7416] to HIRA/HIR - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HIRA/HIR
Clonality: Monoclonal
Clone: EPR7416
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB232477-100U
- ABCA_AB232477-1MG
- Antigen symbol:
- HIRA/HIR
- HIRA/HIR
- Antigen name:
- DGCR1
- DGCR1
- Antigen synonyms:
- HIR|TUPLE1|HIRA|Protein HIRA|TUP1-like enhancer of split protein 1
- HIR|TUPLE1|HIRA|Protein HIRA|TUP1-like enhancer of split protein 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR7416
- EPR7416
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular weight:
- 111835 Da
- 111835 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
Specifications
Frequently Bought Together