Anti-FeLV p27 Mouse Monoclonal Antibody [clone: PF12J-10A]
Catalog # ABCA_AB46531-125UG
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:FPS
- Antigen symbol:FeLV p27
- Clonality:Monoclonal
- Clone:PF12J-10A
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Size:125 µg
- Form:Liquid
- Antigen synonyms:Feline leukemia provirus|Feline leukemia virus p27|Feline leukemia virus subgroup c receptor related protein 1|Feline sarcoma virus|Oncogene fes|hflvcr
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping temperature:4 °C
- Tested applications:IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [PF12J-10A] to FeLV p27.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FeLV p27
Clonality: Monoclonal
Clone: PF12J-10A
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Feline leukemia virus