Anti-ANK-3 Mouse Monoclonal Antibody [clone: 2A8]
Catalog # ABCA_AB315200-100U
Supplier: Abcam
New Product
Specifications
- Size:100 µl
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [2A8] to ANK-3.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: ANK-3
Clonality: Monoclonal
Clone: 2A8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat;Human