Anti-RAB11FIP1 Rabbit Polyclonal Antibody
Catalog # ABCA_AB251756-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:RCP
- Antigen symbol:RAB11FIP1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen synonyms:Rab11 family-interacting protein 1|Rab11-FIP1|Rab-coupling protein
- Molecular weight:137167 Da
- Storage temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipping temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human RAB11FIP1 aa 750-850. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to RAB11FIP1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RAB11FIP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human