Anti-Staphylococcus aureus Rabbit Polyclonal Antibody (Biotinylated)
Catalog # ABCA_AB68954-500UL
Supplier: Abcam
New Product
Specifications
- Size:500 µl
- Packaging:Vial
Specifications
About this item
Biotin Rabbit polyclonal to Staphylococcus aureus.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Staphylococcus aureus
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. aureus