Anti-TPR Rabbit Polyclonal Antibody
Catalog # ABCA_AB84516-100UL
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:Nucleoprotein TPR
- Antigen symbol:TPR
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen synonyms:Megator|NPC-associated intranuclear protein|Translocated promoter region protein
- Storage buffer:BSA, Tris buffered saline
- Molecular weight:267293 Da
- Storage temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human Nucleoprotein TPR aa 2300 - 2350. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to TPR.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TPR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human