Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: OX26]
Catalog # ABCA_AB244555-100U
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD71
- Antigen symbol:Transferrin Receptor
- Clonality:Monoclonal
- Clone:OX26
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Size:100 µg
- Antigen synonyms:Transferrin receptor protein 1|TR|TfR|TfR1|Trfr|T9|p90|TFRC
- Molecular weight:84871 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at 4 °C. Do not freeze.
- Shipping temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested applications:IHC-P
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [OX26] to Transferrin Receptor - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Transferrin Receptor
Clonality: Monoclonal
Clone: OX26
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat;Mouse