Anti-Cytochrome P450 1A2 Mouse Monoclonal Antibody [clone: d15 (16VII F10F12)]
Catalog # ABCA_AB22717-100UG
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cyp1a-2
- Antigen symbol:Cytochrome P450 1A2
- Clonality:Monoclonal
- Clone:d15 (16VII F10F12)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Cyp1a2|CYPIA2|Cholesterol 25-hydroxylase|Cytochrome P-448|Cytochrome P-450d|Cytochrome P450-D|Hydroperoxy icosatetraenoate dehydratase
- Storage buffer:PBS
- Molecular weight:58259 Da
- Storage temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1.49 mg/ml
- Shipping temperature:Shipped at +4 °C
- Immunogen:Full Length Protein corresponding to Rat Cyp1a2. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF, IHC-P
- Purification:Protein A/G Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [d15 (16VII F10F12)] to Cytochrome P450 1A2.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cytochrome P450 1A2
Clonality: Monoclonal
Clone: d15 (16VII F10F12)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat